Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human/Mouse/Rat NDFIP1 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF8246
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF8246 100 μg
AF8246SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. AF8246 Supplier R&D Systems Supplier No. AF8246
Add to Cart
Add to Cart
This item is not returnable. View return policy

Sheep Polyclonal Antibody

NDFIP1 Polyclonal specifically detects NDFIP1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry.

Specifications

Antigen NDFIP1
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 2 ug/mL, Immunocytochemistry 5-15 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. Q9BT67
Gene Alias Breast cancer-associated protein SGA-1M, MGC10924, N4WBP5, N4WBP5Putative NF-kappa-B-activating protein 164, Nedd4 family interacting protein 1, NEDD4 family-interacting protein 1, NEDD4 WW domain-binding protein 5, Putative MAPK-activating protein PM13, Putative NFKB and MAPK-activating protein
Gene Symbols NDFIP1
Host Species Sheep
Immunogen E. coli-derived recombinant human NDFIP1 Ala10-Asp105 Accession # Q9BT67
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 80762
Test Specificity Detects human, mouse and rat NDFIP1 in Western blots.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Human, Mouse, Rat
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.