Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human/Mouse/Rat Neurogranin Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems MAB7947
This item is not returnable.
View return policy
Description
Neurogranin Monoclonal specifically detects Neurogranin in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.Specifications
Neurogranin | |
Monoclonal | |
LYOPH | |
Western Blot 2 ug/mL, Immunohistochemistry 8-25 ug/mL | |
calmodulin-binding protein, hng, neurogranin, neurogranin (protein kinase C substrate, RC3), Ng, NRGN, protein kinase C substrate, RC3, RC3ng | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
Primary | |
Detects human Neurogranin in ELISAs. | |
Human, Mouse, Rat | |
Purified |
Western Blot, Immunohistochemistry | |
898502 | |
Unconjugated | |
Q92686 | |
NRGN | |
E. coli-derived recombinant human Neurogranin Met1-Asp78 Accession # Q92686 | |
100 μg | |
4900 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG3 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction