Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human/Mouse/Rat Neurogranin Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF7947
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF7947 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AF7947 Supplier R&D Systems Supplier No. AF7947
Add to Cart
Add to Cart
This item is not returnable. View return policy

Sheep Polyclonal Antibody

Neurogranin Polyclonal specifically detects Neurogranin in Human, Mouse, Rat samples. It is validated for Western Blot, Simple Western, Immunohistochemistry.

Specifications

Antigen Neurogranin
Applications Western Blot, Immunohistochemistry
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 0.5 ug/mL, Simple Western 20 ug/mL, Immunohistochemistry 5-15 ug/mL
Gene Accession No. Q92686
Gene Alias calmodulin-binding protein, hng, neurogranin, neurogranin (protein kinase C substrate, RC3), Ng, NRGN, protein kinase C substrate, RC3, RC3ng
Gene Symbols NRGN
Host Species Sheep
Immunogen E. coli-derived recombinant human Neurogranin Met1-Asp78 Accession # Q92686
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4900
Test Specificity Detects human, mouse, and rat Neurogranin Western blots.
Reconstitution Sterile PBS to a final concentration of 0.2 mg/mL.
Target Species Human, Mouse, Rat
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.