Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human/Mouse/Rat NF-L Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems MAB22163100
This item is not returnable.
View return policy
Description
NF-L Monoclonal specifically detects NF-L in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.Specifications
NF-L | |
Monoclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
Mouse | |
100 μg | |
4747 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. |
Western Blot, Immunohistochemistry | |
1002615 | |
Western Blot 2 μ/mL, Immunohistochemistry 1-25 μ/mL | |
68 kDa neurofilament protein, CMT2E, NEFL, neurofilament protein, light chain, neurofilament subunit NF-L, Neurofilament triplet L protein, neurofilament, light polypeptide, neurofilament-light, NF68, NF68FLJ53642, NFL, NF-L, NFLlight polypeptide 68kDa | |
68kD purified NF-L from Bovine spinal cord. | |
Primary | |
Detects human NF-L in direct ELISAs. Detects human, mouse, and rat NF-L in Western blots. | |
Human, Mouse, Rat | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction