Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human/Mouse/Rat NFATC3 Antibody, R&D Systems™

Goat Polyclonal Antibody
Supplier: R&D Systems AF5834SP
This item is not returnable.
View return policy
Description
NFATC3/NFAT4 Polyclonal specifically detects NFATC3/NFAT4 in Human, Mouse, Rat samples. It is validated for Western Blot.Specifications
NFATC3/NFAT4 | |
Polyclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
NFAT4, NF-AT4, NFAT4nuclear factor of activated T-cells, cytoplasmic 3, NFATc3, NF-ATc3, NFATx, nuclear factor of activated T-cells c3 isoform IE-Xa, nuclear factor of activated T-cells, cytoplasmic, calcineurin-dependent 3, T cell transcription factor NFAT4, T-cell transcription factor NFAT4 | |
Goat | |
Affinity Purified | |
RUO | |
4775 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Western Blot | |
LYOPH | |
Western Blot 1 ug/mL | |
Q12968 | |
NFATC3 | |
E. coli-derived recombinant human NFATC3 Asp900-Asp1035 Accession # Q12968 | |
25 μg | |
Primary | |
Detects human, mouse and rat NFATC3 in Western blots. | |
Human, Mouse, Rat | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction