Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human/Mouse/Rat Niemann-Pick Type C1/NPC1 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB10105100
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB10105100 100 μg
MAB10105SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. MAB10105100 Supplier R&D Systems Supplier No. MAB10105100
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Monoclonal Antibody

Niemann-Pick C1 Monoclonal specifically detects Niemann-Pick C1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Knockout Validated.

Specifications

Antigen Niemann-Pick C1
Applications Western Blot, Immunohistochemistry, Immunoassay
Classification Monoclonal
Clone 1318A
Conjugate Unconjugated
Dilution Western Blot 0.5 μg/mL, Immunohistochemistry 3-25 μg/mL, Knockout Validated
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS.
Gene Alias FLJ98532, Niemann-Pick C1 protein, Niemann-Pick disease, type C1, NPC, NPC1, SLC65A1
Host Species Rabbit
Immunogen Synthetic peptide containing Human Niemann-Pick Type C1/NPC1
Purification Method Protein A or G purified from cell culture supernatant
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) 4864
Test Specificity Detects human Niemann-Pick Type C1/NPC1 in direct ELISAs. Detects human, mouse, and rat Niemann-Pick Type C1/NPC1 in Western blots.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human, Mouse, Rat
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution.
Form Supernatant
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.