Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human/Mouse/Rat NKX2.2 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems MAB8167
This item is not returnable.
View return policy
Description
NKX2.2 Monoclonal specifically detects NKX2.2 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry.Specifications
NKX2.2 | |
Monoclonal | |
LYOPH | |
Western Blot 2 ug/mL, Immunocytochemistry 8-25 ug/mL | |
O95096 | |
NKX2-2 | |
E. coli-derived recombinant human NKX2.2 Met1-Lys128 Accession # O95096 | |
100 μg | |
Primary | |
Detects human NKX2.2 in ELISA and Western Blot. | |
Human, Mouse, Rat | |
Purified |
Western Blot, Immunocytochemistry | |
883411 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
Homeobox protein NK-2 homolog B, NK-2 (Drosophila) homolog B, NK2 homeobox 2, NK2 transcription factor related, locus 2, NK2 transcription factor related, locus 2 (Drosophila), NK2 transcription factor-like protein B, Nkx2.2, NKX2.2homeobox protein Nkx-2.2, NKX2B, NKX2BNK-2 homolog B | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
4821 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction