Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human/Mouse/Rat Nrf1 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB5306SP
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB5306SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. MAB5306SP Supplier R&D Systems Supplier No. MAB5306SP
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

Nrf1 Monoclonal specifically detects Nrf1 in Human, Mouse, Rat samples. It is validated for Western Blot.

Specifications

Antigen Nrf1
Applications Western Blot
Classification Monoclonal
Clone 492413
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 2 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. AAH10623
Gene Alias Alpha palindromic-binding protein, Alpha-pal, EWG, HBZ17, LCR-F1, NFE2L1, NRF-1, nuclear respiratory factor 1, TCF11
Gene Symbols NRF1
Host Species Mouse
Immunogen E. coli-derived recombinant human Nrf1 Met1-Lys742 Accession # AAH10623
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4899
Test Specificity Detects endogenous human, mouse and rat Nrf1 at 85 kDa in Western blots.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human, Mouse, Rat
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2a
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.