Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human/Mouse/Rat Nucleostemin Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 5 publications
Supplier: R&D Systems AF1638SP
This item is not returnable.
View return policy
Description
Nucleostemin Polyclonal specifically detects Nucleostemin in Human, Mouse, Rat samples. It is validated for Western Blot, Simple Western, Immunocytochemistry.Specifications
Nucleostemin | |
Polyclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
C77032, E2IG3NNP47, estradiol-induced nucleotide binding protein, GNL3, guanine nucleotide binding protein-like 3 (nucleolar), guanine nucleotide-binding protein-like 3, MGC800, Novel nucleolar protein 47, NSE2-induced gene 3 protein, Nucleolar GTP-binding protein 3, nucleostemin | |
Goat | |
Affinity Purified | |
RUO | |
26354 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Western Blot, Immunocytochemistry | |
LYOPH | |
Western Blot 0.25 ug/mL, Simple Western 5 ug/mL, Immunocytochemistry 5-15 ug/mL | |
Q811S9 | |
GNL3 | |
E. coli-derived recombinant rat Nucleostemin Lys2-Ile538 Accession # Q811S9 | |
25 μg | |
Primary | |
Detects human, mouse, rat Nucleostemin in direct ELISAs and Western blots. In direct ELISAs, less than 1% cross-reactivity with recombinant rat Nucleostemin C-terminal peptide (aa 281-538) is observed. Rat Nucleostemin specific IgG was purified by first passing the sera over a rat Nucleostemin aa 2-538 column and then passing the bound fraction over a rat Nucleostemin aa 281-538 column to removed rat Nucleostemin C-terminal specific IgG. | |
Human, Mouse, Rat | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction