Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human/Mouse/Rat Nucleostemin Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF1638
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF1638 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AF1638 Supplier R&D Systems Supplier No. AF1638
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody has been used in 5 publications

Nucleostemin Polyclonal specifically detects Nucleostemin in Human, Mouse, Rat samples. It is validated for Western Blot, Simple Western, Immunocytochemistry.

Specifications

Antigen Nucleostemin
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 0.25 ug/mL, Simple Western 5 ug/mL, Immunocytochemistry 5-15 ug/mL
Gene Accession No. Q811S9
Gene Alias C77032, E2IG3NNP47, estradiol-induced nucleotide binding protein, GNL3, guanine nucleotide binding protein-like 3 (nucleolar), guanine nucleotide-binding protein-like 3, MGC800, Novel nucleolar protein 47, NSE2-induced gene 3 protein, Nucleolar GTP-binding protein 3, nucleostemin
Gene Symbols GNL3
Host Species Goat
Immunogen E. coli-derived recombinant rat Nucleostemin Lys2-Ile538 Accession # Q811S9
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 26354
Test Specificity Detects human, mouse, rat Nucleostemin in direct ELISAs and Western blots. In direct ELISAs, less than 1% cross-reactivity with recombinant rat Nucleostemin C-terminal peptide (aa 281-538) is observed. Rat Nucleostemin specific IgG was purified by first passing the sera over a rat Nucleostemin aa 2-538 column and then passing the bound fraction over a rat Nucleostemin aa 281-538 column to removed rat Nucleostemin C-terminal specific IgG.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Human, Mouse, Rat
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.