Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human/Mouse/Rat p27/Kip1 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB22561SP
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB22561SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. MAB22561SP Supplier R&D Systems Supplier No. MAB22561SP
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rat Monoclonal Antibody

p27/Kip1 Monoclonal specifically detects p27/Kip1 in Human, Mouse, Rat samples. It is validated for Immunohistochemistry.

Specifications

Antigen p27/Kip1
Applications Immunohistochemistry
Classification Monoclonal
Clone 341116
Concentration LYOPH
Conjugate Unconjugated
Dilution Immunohistochemistry 8-25 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. P46527
Gene Alias CDKN1B, CDKN4, cyclin-dependent kinase inhibitor 1B, cyclin-dependent kinase inhibitor 1B (p27, Kip1), Cyclin-dependent kinase inhibitor p27, Kip1, KIP1P27KIP1, MEN1B, MEN4, p27Kip1
Gene Symbols CDKN1B
Host Species Rat
Immunogen E. coli-derived recombinant human p27/Kip1 Met1-Thr198 Accession # P46527
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1027
Test Specificity Detects human p27/Kip1 in direct ELISAs. Specificity was demonstrated by comparing staining on tissue sections of normal versus p27/Kip1 knockout mice.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human, Mouse, Rat
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2a
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.