Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human/Mouse/Rat p38 alpha Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF8691SP
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF8691SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AF8691SP Supplier R&D Systems Supplier No. AF8691SP
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody has been used in 5 publications

p38 alpha Polyclonal specifically detects p38 alpha in Human, Mouse, Rat samples. It is validated for Western Blot, Simple Western, Immunohistochemistry, Knockout Validated.

Specifications

Antigen p38 alpha
Applications Western Blot, Immunohistochemistry, KnockDown
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 0.5 ug/mL, Simple Western 5 ug/mL, Immunohistochemistry 5-15 ug/mL, Knockout Validated
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. Q16539
Gene Symbols MAPK14
Host Species Rabbit
Immunogen E. coli-derived recombinant human p38 alpha Met1-Ser360 Accession # Q16539
Purification Method Affinity Purified
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1432
Test Specificity Detects human, mouse and rat p38 alpha . Does not detect recombinant human p38 beta , p38 gamma or p38 delta .
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Human, Mouse, Rat
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.