Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human/Mouse/Rat p53 Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 5 publications
Supplier: R&D Systems MAB1355SP
This item is not returnable.
View return policy
Description
p53 Monoclonal specifically detects p53 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunoprecipitation, Immunocytochemistry.Specifications
p53 | |
Monoclonal | |
LYOPH | |
Western Blot 1 ug/mL, Immunoprecipitation 1-2 ug/500 ug cell lysate, Immunocytochemistry 8-25 ug/mL | |
P04637 | |
TP53 | |
E. coli-derived recombinant human p53 Asp7-Asp393 Accession # P04637 | |
25 μg | |
Primary | |
Detects human, mouse, and rat p53. | |
Human, Mouse, Rat | |
Purified |
Western Blot, Immunoprecipitation, Immunocytochemistry | |
184721 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
Antigen NY-CO-13, BCC7, FLJ92943, LFS1, LFS1TRP53, p53, p53 tumor suppressor, P53cellular tumor antigen p53, Phosphoprotein p53, TP53, transformation-related protein 53, TRP53, tumor protein p53, Tumor suppressor p53 | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
7157 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction