Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human/Mouse/Rat p53 Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 12 publications
Supplier: R&D Systems AF1355
This item is not returnable.
View return policy
Description
p53 Polyclonal specifically detects p53 in Human, Mouse, Rat samples. It is validated for Western Blot, Simple Western, Immunoprecipitation, Chromatin Immunoprecipitation (ChIP), Immunocytochemistry, Knockout Validated.Specifications
p53 | |
Polyclonal | |
Unconjugated | |
P04637 | |
TP53 | |
E. coli-derived recombinant human p53 Asp7-Asp393 Accession # P04637 | |
100 μg | |
Primary | |
Detects human, mouse, and rat p53. | |
Human, Mouse, Rat | |
IgG |
ChIP Assay, Immunocytochemistry, Immunoprecipitation | |
LYOPH | |
Western Blot 0.25 ug/mL, Simple Western 2.5 ug/mL, Immunoprecipitation 1-2 ug/500 ug cell lysate, Chromatin Immunoprecipitation (ChIP) 5 ug/5 x 10^6 cells, Immunocytochemistry 5-15 ug/mL, Knockout Validated | |
Antigen NY-CO-13, BCC7, FLJ92943, LFS1, LFS1TRP53, p53, p53 tumor suppressor, P53cellular tumor antigen p53, Phosphoprotein p53, TP53, transformation-related protein 53, TRP53, tumor protein p53, Tumor suppressor p53 | |
Goat | |
Affinity Purified | |
RUO | |
7157 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction