Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human/Mouse/Rat p53 HRP-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. HAF1355
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
HAF1355 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. HAF1355 Supplier R&D Systems Supplier No. HAF1355
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody has been used in 9 publications

p53 Polyclonal specifically detects p53 in Human, Mouse, Rat samples. It is validated for Western Blot.

Specifications

Antigen p53
Applications Western Blot
Classification Polyclonal
Conjugate HRP
Dilution Western Blot 1:5000 dilution
Gene Accession No. P04637
Gene Alias Antigen NY-CO-13, BCC7, FLJ92943, LFS1, LFS1TRP53, p53, p53 tumor suppressor, P53cellular tumor antigen p53, Phosphoprotein p53, TP53, transformation-related protein 53, TRP53, tumor protein p53, Tumor suppressor p53
Gene Symbols TP53
Host Species Goat
Immunogen E. coli-derived recombinant human p53 Asp7-Asp393 Accession # P04637
Purification Method Affinity Purified
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7157
Test Specificity Detects human, mouse, and rat p53 in Western blots.
Target Species Human, Mouse, Rat
Content And Storage Do not freeze. 6 months from date of receipt, 2 to 8 degreesC as supplied.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.