Learn More
Human/Mouse/Rat p62/SQSTM1 Antibody, R&D Systems™

Description
Specifications
Specifications
| Antigen | p62/SQSTM1 |
| Applications | Western Blot, Immunohistochemistry, Immunoprecipitation, Immunocytochemistry, Immunoassay |
| Classification | Monoclonal |
| Clone | 864807 |
| Conjugate | Unconjugated |
| Dilution | Western Blot 2 ug/mL, Simple Western 20 ug/mL, Immunohistochemistry 5-25 ug/mL, Immunoprecipitation 1 ug/1 mg cell lysate, Immunocytochemistry 8-25 ug/mL, Knockout Validated |
| Formulation | Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative |
| Gene Accession No. | Q13501 |
| Gene Alias | A170, Autophagy Receptor P62, DMRV, EBI3-associated protein of 60 kDa, EBI3-associated protein p60, EBIAP, FTDALS3, NADGP, ORCA, OSIL, oxidative stress induced like, p60PDB3, p62, p62B, Paget disease of bone 3, PDB3, phosphotyrosine independent ligand for the Lck SH2 domain p62, Phosphotyrosine-independent ligand for the Lck SH2 domain of 62 kDa, sequestosome 1, sequestosome-1, SQSTM1, Ubiquitin-binding protein p62, ZIP3 |
| Gene Symbols | SQSTM1 |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.