Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human/Mouse/Rat p62/SQSTM1 Biotinylated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. BAM8028
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BAM8028 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. BAM8028 Supplier R&D Systems Supplier No. BAM8028
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

p62/SQSTM1 Monoclonal specifically detects p62/SQSTM1 in Human, Mouse, Rat samples. It is validated for Immunocytochemistry.

Specifications

Antigen p62/SQSTM1
Applications Immunocytochemistry
Classification Monoclonal
Clone 864807
Concentration LYOPH
Conjugate Biotin
Dilution Immunocytochemistry 8 ug/mL
Gene Accession No. Q13501
Gene Alias A170, Autophagy Receptor P62, DMRV, EBI3-associated protein of 60 kDa, EBI3-associated protein p60, EBIAP, FTDALS3, NADGP, ORCA, OSIL, oxidative stress induced like, p60PDB3, p62, p62B, Paget disease of bone 3, PDB3, phosphotyrosine independent ligand for the Lck SH2 domain p62, Phosphotyrosine-independent ligand for the Lck SH2 domain of 62 kDa, sequestosome 1, sequestosome-1, SQSTM1, Ubiquitin-binding protein p62, ZIP3
Gene Symbols SQSTM1
Host Species Mouse
Immunogen E. coli-derived recombinant human p62/SQSTM1 Asp368-Leu440 Accession # Q13501
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 50 μg
Primary or Secondary Primary
Gene ID (Entrez) 8878
Test Specificity p62/SQSTM1 antibodies are ideal for immunocytochemistry colocalization studies in autophagosomes. The unconjugated antibody detects human p62/SQSTM1 in direct ELISAs, and human, mouse and rat p62/SQSTM1 in Western Blots
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human, Mouse, Rat
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.