Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human/Mouse/Rat PALS1/MPP5 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF7979
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF7979 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AF7979 Supplier R&D Systems Supplier No. AF7979
Add to Cart
Add to Cart
This item is not returnable. View return policy

Sheep Polyclonal Antibody

PALS1/MPP5 Polyclonal specifically detects PALS1/MPP5 in Human, Mouse, Rat samples. It is validated for Western Blot.

Specifications

Antigen PALS1/MPP5
Applications Western Blot
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 0.5 ug/mL
Gene Accession No. Q8N3R9
Gene Alias MAGUK p55 subfamily member 5, membrane protein, palmitoylated 5 (MAGUK p55 subfamily member 5), MPP5, PALS1FLJ12615, stardust
Gene Symbols MPP5
Host Species Sheep
Immunogen E. coli-derived recombinant human PALS1/MPP5 Ala392-Ser509 Accession # Q8N3R9
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 64398
Test Specificity Detects human, mouse, and rat PALS1/MPP5 in direct ELISAs and Western blots.
Reconstitution Sterile PBS to a final concentration of 0.2 mg/mL.
Target Species Human, Mouse, Rat
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.