Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human/Mouse/Rat Pannexin-1 Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 4 publications
Supplier: R&D Systems MAB7097
This item is not returnable.
View return policy
Description
Pannexin-1 Monoclonal specifically detects Pannexin-1 in Human, Mouse, Rat samples. It is validated for Western Blot.Specifications
Pannexin-1 | |
Monoclonal | |
LYOPH | |
Western Blot 1 ug/mL | |
Q96RD7 | |
PANX1 | |
E. coli-derived recombinant human Pannexin-1 Pro298-Asp376 Accession # Q96RD7 | |
100 μg | |
Primary | |
Detects human Pannexin-1 in direct ELISAs and Western blots. In Western blots, no cross-reactivity with recombinant human Pannexin-2 or -3 is observed. | |
Human, Mouse, Rat | |
Purified |
Western Blot | |
720505 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
Innexin, MGC21309, MRS1, MRS1innexin, pannexin 1, pannexin-1, Pannexin1, PANX1, PX1, UNQ2529 | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
24145 | |
Sterile PBS to a final concentration of 0.5 mg/mL. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction