Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human/Mouse/Rat Park7/DJ-1 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB39951SP
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB39951SP 25 μg
MAB39951 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. MAB39951SP Supplier R&D Systems Supplier No. MAB39951SP
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

Park7/DJ-1 Monoclonal specifically detects Park7/DJ-1 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunocytochemistry, Knockout Validated.

Specifications

Antigen Park7/DJ-1
Applications Western Blot, ELISA, Immunocytochemistry, KnockDown
Classification Monoclonal
Clone 925805R
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL, ELISA, Immunocytochemistry 8-25 ug/mL, Knockout Validated
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. Q99497
Gene Alias DJ1, DJ-1, DJ1FLJ34360, EC 3.4, FLJ27376, FLJ92274, Oncogene DJ1, Parkinson disease (autosomal recessive, early onset) 7, Parkinson disease protein 7, protein DJ-1
Gene Symbols PARK7
Host Species Mouse
Immunogen E. coli-derived recombinant human PARK-7 Ala2-Asp189 Accession # Q99497
Purification Method Protein A or G purified from cell culture supernatant
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11315
Test Specificity Detects human, mouse, and rat PARK-7 in Western blots. In sandwich immunoassays, it is specific for human PARK-7.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human, Mouse, Rat
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2a
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.