Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human/Mouse/Rat Parvalbumin alpha Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF5058
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF5058 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AF5058 Supplier R&D Systems Supplier No. AF5058
Add to Cart
Add to Cart
This item is not returnable. View return policy

Sheep Polyclonal Antibody has been used in 8 publications

Parvalbumin alpha Polyclonal specifically detects Parvalbumin alpha in Human, Mouse, Rat samples. It is validated for Western Blot, Simple Western, Immunohistochemistry.

Specifications

Antigen Parvalbumin alpha
Applications Western Blot, Immunohistochemistry
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL, Simple Western 50 ug/mL, Immunohistochemistry 5-15 ug/mL
Gene Accession No. P20472
Gene Alias D22S749, PVALB
Gene Symbols PVALB
Host Species Sheep
Immunogen E. coli-derived recombinant human Parvalbumin alpha Ser2-Ser110 Accession # P20472
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5816
Test Specificity Detects human, mouse, and rat Parvalbumin alpha in direct ELISAs and Western blots.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Human, Mouse, Rat
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.