Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human/Mouse/Rat PCK1 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF7316
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF7316 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AF7316 Supplier R&D Systems Supplier No. AF7316
Add to Cart
Add to Cart
This item is not returnable. View return policy

Sheep Polyclonal Antibody

PCK1 Polyclonal specifically detects PCK1 in Human, Mouse, Rat samples. It is validated for Western Blot.

Specifications

Antigen PCK1
Applications Western Blot
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL
Gene Accession No. P35558
Gene Alias EC 4.1.1.32, MGC22652, PEPCK1, PEPCKC, PEPCK-C, PEPCK-CPEP carboxykinase, phosphoenolpyruvate carboxykinase 1 (soluble), phosphoenolpyruvate carboxykinase, cytosolic, phosphoenolpyruvate carboxykinase, cytosolic [GTP], Phosphoenolpyruvate carboxylase, phosphopyruvate carboxylase
Gene Symbols PCK1
Host Species Sheep
Immunogen E. coli-derived recombinant human PCK1 Met1-Ile88 Accession # P35558
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5105
Test Specificity Detects human, mouse, rat PCK1 in Western blots, and detects recombinant human PCK1 in direct ELISAs. In direct ELISAs, less than 3% cross-reactivity with recombinant mouse PCK1 is observed.
Reconstitution Sterile PBS to a final concentration of 0.2 mg/mL.
Target Species Human, Mouse, Rat
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.