Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human/Mouse/Rat PDCD6 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB6015
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB6015 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. MAB6015 Supplier R&D Systems Supplier No. MAB6015
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

PDCD6 Monoclonal specifically detects PDCD6 in Human, Mouse, Rat samples. It is validated for Western Blot.

Specifications

Antigen PDCD6
Applications Western Blot
Classification Monoclonal
Clone 722232
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 0.2 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. O75340
Gene Alias ALG2, ALG-2, Apoptosis-linked gene 2 protein, apoptosis-linked gene-2, FLJ46208, MGC111017, MGC119050, MGC9123, PEF1B, Probable calcium-binding protein ALG-2, programmed cell death 6, programmed cell death protein 6
Gene Symbols PDCD6
Host Species Mouse
Immunogen E. coli-derived recombinant human PDCD6 Met1-Val189 Accession # O75340
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10016
Test Specificity Detects human PDCD6 in direct ELISAs and Western blots. In direct ELISAs, no cross-reactivity with recombinant human (rh) PDCD4 or rhPDCD5 is observed.
Reconstitution Sterile PBS to a final concentration of 0.5 mg/mL.
Target Species Human, Mouse, Rat
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.