Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human/Mouse/Rat Peroxiredoxin 2 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF3489
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF3489 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AF3489 Supplier R&D Systems Supplier No. AF3489
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody has been used in 3 publications

Peroxiredoxin 2 Polyclonal specifically detects Peroxiredoxin 2 in Human, Mouse, Rat samples. It is validated for Western Blot, Simple Western.

Specifications

Antigen Peroxiredoxin 2
Applications Western Blot
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 0.2 ug/mL, Simple Western 2 ug/mL
Gene Accession No. P32119
Gene Alias EC 1.11.1, MGC4104, Natural killer cell-enhancing factor B, natural killer-enhancing factor B, NKEF-B, NKEFBNKEF-B, peroxiredoxin 2, peroxiredoxin-2, PRDX2, PRPTDPX1, PRX2, PRXII, TDPX1, thiol-specific antioxidant 1, Thiol-specific antioxidant protein, Thioredoxin peroxidase 1, Thioredoxin-dependent peroxide reductase 1, torin, TPX1, TSA, TSAEC 1.11.1.15
Gene Symbols PRDX2
Host Species Goat
Immunogen E. coli-derived recombinant human Peroxiredoxin 2 Met1-Asn198 Accession # P32119
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7001
Test Specificity The antibody detects endogenous human, mouse and rat Peroxiredoxin 2 in in Western blots. In Western blots, approximately 25% cross-reactivity with recombinant human (rh) Peroxiredoxin 1 and rhPeroxiredoxin 4 is observed, approximately 10% cross-reactivity with rhPeroxiredoxin 3 is observed, and less than 1% cross-reactivity with rhPeroxiredoxin 5 and rhPeroxiredoxin 6 is observed.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Human, Mouse, Rat
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.