Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human/Mouse/Rat PGK1 Antibody, R&D Systems™

Goat Polyclonal Antibody
Supplier: R&D Systems AF5455
This item is not returnable.
View return policy
Description
PGK1 Polyclonal specifically detects PGK1 in Human, Mouse, Rat samples. It is validated for Western Blot.Specifications
PGK1 | |
Polyclonal | |
Unconjugated | |
P00558 | |
PGK1 | |
S. frugiperda insect ovarian cell line Sf 21-derived recombinant human PGK1 Met1-Ile417 Accession # P00558 | |
100 μg | |
Primary | |
Detects endogenous human, mouse and rat PGK1 in Western blots. | |
Human, Mouse, Rat | |
IgG |
Western Blot | |
LYOPH | |
Western Blot 1 ug/mL | |
Cell migration-inducing gene 10 protein, EC 2.7.2.3, MGC142128, MGC8947, MIG10, PGKA, PGKAMGC117307, phosphoglycerate kinase 1, Primer recognition protein 2, PRP 2, PRP2 | |
Goat | |
Affinity Purified | |
RUO | |
5230 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction