Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human/Mouse/Rat Phospho-CDC2/CDK1 (Y15) Antibody, R&D Systems™
Rabbit Polyclonal Antibody has been used in 3 publications
Supplier: R&D Systems AF888
This item is not returnable.
View return policy
Description
CDC2/CDK1 Polyclonal specifically detects CDC2/CDK1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry.Specifications
CDC2/CDK1 (p tyr15) | |
Polyclonal | |
Unconjugated | |
CDC28A, CDC2MGC111195, CDK1, cell cycle controller CDC2, Cell division control protein 2 homolog, Cell division protein kinase 1, cyclin-dependent kinase 1, DKFZp686L20222, EC 2.7.11.22, EC 2.7.11.23, G1 to S and G2 to M, p34 protein kinase, P34CDC2 | |
Rabbit | |
Affinity Purified | |
RUO | |
983 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Western Blot, Immunocytochemistry | |
LYOPH | |
Western Blot 0.2 ug/mL, Immunocytochemistry 5-15 ug/mL | |
CDK1 | |
Phosphopeptide containing CDC2 Y15 site | |
100 μg | |
Primary | |
Detects human, mouse, and rat CDC2 when phosphorylated at Y15. | |
Human, Mouse, Rat | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction