Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human/Mouse/Rat Phospho-Chk1 (S317) Antibody, R&D Systems™

Rabbit Polyclonal Antibody has been used in 7 publications
Supplier: R&D Systems AF2054
This item is not returnable.
View return policy
Description
Chk1 Polyclonal specifically detects Chk1 in Human, Mouse, Rat samples. It is validated for Western Blot, Simple Western.Specifications
Chk1 (p Ser317) | |
Polyclonal | |
Unconjugated | |
Checkpoint, S. pombe, homolog of, 1, CHEK1, CHK1 (checkpoint, S.pombe) homolog, CHK1 checkpoint homolog (S. pombe), CHK1serine/threonine-protein kinase Chk1, EC 2.7.11, EC 2.7.11.1 | |
Rabbit | |
Affinity Purified | |
RUO | |
1111 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Western Blot | |
LYOPH | |
Western Blot 1 ug/mL, Simple Western 10 ug/mL | |
CHEK1 | |
Phosphopeptide containing human Chk1 S317 site | |
100 μg | |
Primary | |
Detects human, mouse, and rat Chk1 when phosphorylated at S317 in Western blots. Does not recognize Chk1 when unphosphorylated at S317. | |
Human, Mouse, Rat | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction