Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human/Mouse/Rat Phospho-ERK1/ERK2 (ERK1 T202/Y204, ERK2 T185/Y187) Alexa Fluor 750-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. IC1018S100
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
IC1018S100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. IC1018S100 Supplier R&D Systems Supplier No. IC1018S100UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Monoclonal Antibody

ERK1/ERK2 Monoclonal specifically detects ERK1/ERK2 in Human, Mouse, Rat samples. It is validated for Intracellular Staining by Flow Cytometry.

Specifications

Antigen ERK1/ERK2
Applications Flow Cytometry
Classification Monoclonal
Clone 269434
Conjugate Alexa Fluor 750
Dilution Intracellular Staining by Flow Cytometry 0.25-1 ug/10^6 cells
Formulation Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide.
Host Species Rabbit
Immunogen Phosphopeptide containing ERK1 T202/Y204 site
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity Detects human, mouse, and rat ERK1 and ERK2 dually phosphorylated at T202/Y204 or T185/Y187, respectively.
Target Species Human, Mouse, Rat
Content And Storage Store the unopened product at 2 - 8 degreesC. Do not use past expiration date.
Isotype IgG
Show More Show Less

For Research Use Only (RUO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.