Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Human/Mouse/Rat Phospho-ERK2 (T185/Y187) DuoSet IC

Catalog No. DYC14832
Change view
Click to view available options
Format:
15 96-well Plates
2 96-well Plates
5 96-well Plates
Gene ID (Entrez):
5594
26413
116590
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Format Gene ID (Entrez)
DYC14832 2 96-well Plates 116590
DYC1483E 15 96-well Plates 5594
DYC14835 5 96-well Plates 26413
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. DYC14832 Supplier R&D Systems™ Supplier No. DYC14832
This item is not returnable. View return policy

Phospho-ERK2 (T185/Y187) DuoSet IC ELISA from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.

The Phospho-ERK2 (T185/Y187) DuoSet IC ELISA is a Solid Phase Sandwich ELISA that quantifies phosphorylated human, mouse, rat ERK2.

Specifications

Product Type ELISA Development Kit for Phosphorylated Intracellular Proteins
Quantity 1 Kit (for 2 Plates)
Species Human, Mouse, Rat
Conjugate Biotin
Assay Range 188.00 to 12000 pg/mL
Cross Reactivity <50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules
Format 2 96-well Plates
For Use With (Application) ELISA
Gene ID (Entrez) 116590
Synonym EC 2.7.11, EC 2.7.11.24, ERK, ERK-2, ERK2MAP kinase isoform p42, ERT1, Extracellular signal-regulated kinase 2, MAP kinase 1, MAP kinase 2, MAPK 1, MAPK1, MAPK2, mitogen-activated protein kinase 1, Mitogen-activated protein kinase 2, p38, p40, p41, p41mapk, p42-MAPK, P42MAPK, PRKM1, PRKM1MAPK 2, PRKM2, protein tyrosine kinase ERK2
Content And Storage Store the unopened product at 2°C to 8°C. Do not use past expiration date.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.