Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human/Mouse/Rat Phospho-HSP27 (S78/S82) Antibody, R&D Systems™

Rabbit Polyclonal Antibody has been used in 2 publications
Supplier: R&D Systems AF2314SP
This item is not returnable.
View return policy
Description
HSP27 Polyclonal specifically detects HSP27 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry.Specifications
HSP27 (p ser82) | |
Polyclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
HSPB1 | |
Phosphopeptide containing human HSP27 S78/S82 sites | |
25 μg | |
Primary | |
Detects human HSP27 when dually phosphorylated at S78/S82, and mouse and rat HSP27 phosphorylated at S86. | |
Human, Mouse, Rat | |
IgG |
Western Blot, Immunocytochemistry | |
LYOPH | |
Western Blot 0.1 ug/mL, Immunocytochemistry 5-15 ug/mL | |
28 kDa heat shock protein, DKFZp586P1322, Estrogen-regulated 24 kDa protein, Heat shock 27 kDa protein, heat shock 27kD protein 1, heat shock 27kDa protein 1, heat shock protein beta-1, HMN2B, HS.76067, Hsp25, HSP27HSP 27, HSP28CMT2F, HSPB1, SRP27, Stress-responsive protein 27 | |
Rabbit | |
Affinity Purified | |
RUO | |
3315 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction