Learn More
R&D Systems™ Human/Mouse/Rat Phospho-JNK Pan Specific DuoSet IC ELISA
Description
Specifications
Specifications
| Product Type | ELISA Kit |
| Quantity | 1 Kit (for 2 Plates) |
| Species | Human, Mouse, Rat |
| Conjugate | Biotin |
| Assay Range | 156.00 to 10000 pg/mL |
| Format | 2 96-well Plates |
| For Use With (Application) | ELISA |
| Synonym | c-Jun N-terminal kinase 1, EC 2.7.11, EC 2.7.11.24, JNK1 alpha protein kinase, JNK1 beta protein kinase, JNK1JNK1A2, JNK-46, JUN N-terminal kinase, MAP kinase 8, MAPK 8, MAPK10, MAPK8, MAPK9, mitogen-activated protein kinase 8, mitogen-activated protein kinase 8 isoform JNK1 alpha1, mitogen-activated protein kinase 8 isoform JNK1 beta2, PRKM8, PRKM8JNK, protein kinase JNK1, SAPK1, SAPK1 alpha, SAPK1 beta, SAPK1JNK21B1/2, Stress-activated protein kinase 1, Stress-activated protein kinase JNK1 |
| Content And Storage | Store the unopened product at 2°C to 8°C. Do not use past expiration date. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.