Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human/Mouse/Rat Phospho-JNK (T183/Y185) Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB1205
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB1205 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. MAB1205 Supplier R&D Systems Supplier No. MAB1205
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Monoclonal Antibody

JNK1/2/3 Monoclonal specifically detects JNK1/2/3 in Human, Mouse, Rat samples. It is validated for Western Blot, Simple Western, Immunocytochemistry.

Order Info

The product is shipped with polar packs.

Specifications

Antigen JNK1/2/3 (p Thr183, p Tyr185)
Applications Western Blot, Immunocytochemistry
Classification Monoclonal
Clone 1006A
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL, Simple Western 10-25 ug/mL, Immunocytochemistry 10-35 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with Sodium Azide
Gene Alias c-Jun N-terminal kinase 1, EC 2.7.11, EC 2.7.11.24, JNK1 alpha protein kinase, JNK1 beta protein kinase, JNK1JNK1A2, JNK-46, JUN N-terminal kinase, MAP kinase 8, MAPK 8, MAPK10, MAPK8, MAPK9, mitogen-activated protein kinase 8, mitogen-activated protein kinase 8 isoform JNK1 alpha1, mitogen-activated protein kinase 8 isoform JNK1 beta2, PRKM8, PRKM8JNK, protein kinase JNK1, SAPK1, SAPK1 alpha, SAPK1 beta, SAPK1JNK21B1/2, Stress-activated protein kinase 1, Stress-activated protein kinase JNK1
Gene Symbols MAPK8IP3
Host Species Rabbit
Immunogen Phosphopeptide containing human, rat, and mouse JNK1 T183/Y185 site
Purification Method Protein A or G purified from cell culture supernatant
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5599
Test Specificity Detects human, mouse and rat p46 and p54 JNK when dually phosphorylated at sites homologous to T183/Y185 of JNK1 and JNK2, and T221/Y223 of JNK3 in Western blots.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human, Mouse, Rat
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.