Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human/Mouse/Rat Phospho-MEK1 (S218/S222)/MEK2 (S222/S226) Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB8407SP
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB8407SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. MAB8407SP Supplier R&D Systems Supplier No. MAB8407SP
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Monoclonal Antibody

MEK1/2 Monoclonal specifically detects MEK1/2 in Human, Mouse, Rat samples. It is validated for Western Blot.

Specifications

Antigen MEK1/2
Applications Western Blot
Classification Monoclonal
Clone 1020E
Conjugate Unconjugated
Dilution Western Blot 0.5 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with Sodium Azide
Gene Alias CFC3, CFC4, dual specificity mitogen-activated protein kinase kinase 1, dual specificity mitogen-activated protein kinase kinase 2, ERK activator kinase 1, ERK activator kinase 2, MAP kinase kinase 2, MAP2K1, MAP2K2, MAPK/ERK kinase 1, MAPK/ERK kinase 2, MAPKK 1, MAPKK1, MAPKK2, MEK 1, MEK1, MEK2, MEL, mitogen-activated protein kinase kinase 2, p45, MKK1, MKK2, PRKMK1, PRKMK2, protein kinase, mitogen-activated, kinase 1 (MAP kinase kinase 1)
Gene Symbols MAP2K1
Host Species Rabbit
Immunogen Phosphopeptide containing the human, mouse, and rat MEK1 S218/S222 site
Purification Method Protein A or G purified from cell culture supernatant
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5604
Test Specificity Detects human, mouse, and rat MEK1 and MEK2 dually phosphorylated at S218/S222 and S222/S226, respectively.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS
Target Species Human, Mouse, Rat
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.