Learn More
Human/Mouse/Rat Phospho-p70 S6 Kinase (T421/S424) Antibody, R&D Systems™
GREENER_CHOICE
Rabbit Polyclonal Antibody has been used in 6 publications
Supplier:  R&D Systems AF8965SP
Catalog No. AF8965SP
Only null left
Add to Cart
This item is not returnable. View return policy
Description
Description
p70 S6 Kinase/S6K Polyclonal specifically detects p70 S6 Kinase/S6K in Human, Mouse, Rat samples. It is validated for Western Blot, Simple Western.
Specifications
Specifications
p70 S6 Kinase/S6K (p Thr421, p Ser424)
Polyclonal
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
RPS6KB1
Phosphopeptide containing human, rat, and mouse p70 S6 Kinase T421/S424 sites
25 μg
Primary
Detects human, mouse and rat p70 S6 Kinase when phosphorylated at T421 and S424 and p85 S6K when phosphorylated at T444 and S447.
Human, Mouse, Rat
IgG
Western Blot
LYOPH
Western Blot 0.25 ug/mL, Simple Western 5 ug/mL
EC 2.7.11, EC 2.7.11.1, p70 ribosomal S6 kinase alpha, p70 S6 kinase alpha, p70 S6 kinase, alpha 1,70 kDa ribosomal protein S6 kinase 1, p70 S6 kinase, alpha 2, p70 S6KA, p70 S6K-alpha, p70(S6K)-alpha, p70-alpha, p70-S6K, P70S6K1, PS6K, ribosomal protein S6 kinase beta-1, Ribosomal protein S6 kinase I, ribosomal protein S6 kinase, 70kD, polypeptide 1, ribosomal protein S6 kinase, 70kDa, polypeptide 1, RPS6KB1, S6K, S6K1, S6K1p70-S6K 1, S6K-beta-1, serine/threonine kinase 14 alpha, Serine/threonine-protein kinase 14A, STK14A
Rabbit
Affinity Purified
RUO
6198
Reconstitute at 0.2 mg/mL in sterile PBS.
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.
For Research Use Only