Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Human/Mouse/Rat Phospho-RPS6 (S235/S236) DuoSet IC ELISA

Catalog No. DYC39185
Change view
Click to view available options
Format:
15 96-well Plates
2 96-well Plates
5 96-well Plates
Gene ID (Entrez):
20104
29304
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Format Gene ID (Entrez)
DYC39185 5 96-well Plates 29304
DYC3918E 15 96-well Plates 20104
DYC39182 2 96-well Plates 29304
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. DYC39185 Supplier R&D Systems™ Supplier No. DYC39185
Add to Cart
Add to Cart
This item is not returnable. View return policy

Contains the basic components required for the development of sandwich ELISAs to measure in cell lysates.

The Human/Mouse/Rat Phospho-RPS6 (S235/S236) DuoSet IC ELISA is a Solid Phase Sandwich ELISA that quantifies phosphorylated human, mouse, rat Ribosomal Protein S6/RPS6.

Specifications

Product Type ELISA Development Kit for Phosphorylated Intracellular Proteins
Quantity 1 Kit
Species Human, Mouse, Rat
Conjugate Biotin
Assay Range 312.50 to 20000 pg/mL
Cross Reactivity <50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules
Format 5 96-well Plates
For Use With (Application) ELISA
Gene ID (Entrez) 29304
Content And Storage Store the unopened product at 2°C to 8°C. Do not use past expiration date.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.