Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human/Mouse/Rat PI 3-Kinase p55 gamma Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems MAB6638SP
This item is not returnable.
View return policy
Description
PI 3-Kinase p55 gamma Monoclonal specifically detects PI 3-Kinase p55 gamma in Human, Mouse, Rat samples. It is validated for Western Blot.Specifications
PI 3-Kinase p55 gamma | |
Monoclonal | |
LYOPH | |
Western Blot 1 ug/mL | |
Q92569 | |
PIK3R3 | |
E. coli-derived recombinant human p55 gamma Lys251-Gly378 Accession # Q92569 | |
25 μg | |
Primary | |
Detects human, mouse, and rat PI 3-Kinase p55 gamma in Western blots. In direct ELISAs, less than 10% cross-reactivity with recombinant human (rh) PIK3R1 (aa 519-644) and no cross-reactivity with rhPIK3R1 (aa 328-431), rhPIK3R2 (aa 325-428), rhPIK3R2 (aa 516-642), rhPIK3C3 (aa 419-887), rhPIK3R4 (aa 58-230), or rhPIK3R5 (aa 530-726) is observed. | |
Human, Mouse, Rat | |
Purified |
Western Blot | |
668619 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
DKFZp686P05226, FLJ41892, p55, p55-GAMMA, p55PIK, Phosphatidylinositol 3-kinase 55 kDa regulatory subunit gamma, phosphatidylinositol 3-kinase regulatory subunit gamma, phosphatidylinositol 3-kinase, regulatory subunit, polypeptide 3 (p55, gamma), phosphoinositide-3-kinase, regulatory subunit 3 (gamma), PI 3Kinase p55 gamma, PI3K regulatory subunit gamma, PI3-kinase regulatory subunit gamma, PI3-kinase subunit p55-gamma, PIK3R3, PtdIns-3-kinase regulatory subunit gamma, ptdIns-3-kinase regulatory subunit p55-gamma, regulatory subunit, polypeptide 3 (p55, gamma) | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
8503 | |
Sterile PBS to a final concentration of 0.5 mg/mL. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction