Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human/Mouse/Rat PI 3-Kinase p55 gamma Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF5734
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF5734 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AF5734 Supplier R&D Systems Supplier No. AF5734
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody

PI 3-Kinase p55 gamma Polyclonal specifically detects PI 3-Kinase p55 gamma in Human, Mouse, Rat samples. It is validated for Western Blot.

Specifications

Antigen PI 3-Kinase p55 gamma
Applications Western Blot
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL
Gene Accession No. Q92569
Gene Alias DKFZp686P05226, FLJ41892, p55, p55-GAMMA, p55PIK, Phosphatidylinositol 3-kinase 55 kDa regulatory subunit gamma, phosphatidylinositol 3-kinase regulatory subunit gamma, phosphatidylinositol 3-kinase, regulatory subunit, polypeptide 3 (p55, gamma), phosphoinositide-3-kinase, regulatory subunit 3 (gamma), PI 3Kinase p55 gamma, PI3K regulatory subunit gamma, PI3-kinase regulatory subunit gamma, PI3-kinase subunit p55-gamma, PIK3R3, PtdIns-3-kinase regulatory subunit gamma, ptdIns-3-kinase regulatory subunit p55-gamma, regulatory subunit, polypeptide 3 (p55, gamma)
Gene Symbols PIK3R3
Host Species Goat
Immunogen E. coli-derived recombinant human PI 3-Kinase p55 gamma Lys251-Gly378 Accession # Q92569
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8503
Test Specificity Detects endogenous human, mouse and rat PI 3-Kinase p55 gamma in Western blots.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Human, Mouse, Rat
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.