Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human/Mouse/Rat PKA C alpha Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF4268SP
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF4268SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AF4268SP Supplier R&D Systems Supplier No. AF4268SP
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody

PKA C alpha Polyclonal specifically detects PKA C alpha in Human, Mouse, Rat samples. It is validated for Western Blot.

Specifications

Antigen PKA C alpha
Applications Western Blot
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 2 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. P17612
Gene Alias cAMP-dependent protein kinase catalytic subunit alpha, Cs, EC 2.7.11, EC 2.7.11.11, MGC102831, MGC48865, PKA C-alpha, PKACA, PKCA1, PKCD, PRKACA, protein kinase A catalytic subunit, protein kinase, cAMP-dependent, catalytic, alpha
Gene Symbols PRKACA
Host Species Goat
Immunogen E. coli-derived recombinant human PKA C alpha Met1-Phe351 Accession # P17612
Purification Method Affinity Purified
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5566
Test Specificity Detects human, mouse, and rat PKA C alpha in Western blots. In Western blots, less than 1% cross-reactivity with recombinant human (rh) PKA C beta or rhPKA C delta is observed.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Human, Mouse, Rat
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.