Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human/Mouse/Rat PKA C Pan Specific Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems MAB4175
This item is not returnable.
View return policy
Description
PKA C (pan) Monoclonal specifically detects PKA C (pan) in Human, Mouse, Rat samples. It is validated for Western Blot.Specifications
PKA C (pan) | |
Monoclonal | |
LYOPH | |
Western Blot 1 ug/mL | |
ABM84853 | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
5566 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG2a |
Western Blot | |
485013 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
CAFD1, cAPKbeta, Cs, Pkaca, PKCA1, PKCD, PPNAD4, PRKACA, PRKACB | |
E. coli-derived recombinant human PKA C gamma Met1-Phe351 Accession # ABM84853 | |
100 μg | |
Primary | |
Detects human PKA C alpha , C beta , and C gamma as well as endogenous human, mouse, and rat PKA C isoforms at 39-42 kDa using Western blot. | |
Human, Mouse, Rat | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction