Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human/Mouse/Rat PKA RI beta Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF4177
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF4177 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AF4177 Supplier R&D Systems Supplier No. AF4177
Add to Cart
Add to Cart
This item is not returnable. View return policy

Sheep Polyclonal Antibody has been used in 2 publications

PKA RI beta Polyclonal specifically detects PKA RI beta in Human, Mouse, Rat samples. It is validated for Western Blot, Simple Western.

Specifications

Antigen PKA RI beta
Applications Western Blot
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL, Simple Western 10 ug/mL
Gene Accession No. P31321
Gene Alias PRKAR1, PRKAR1B, RIbeta
Gene Symbols PRKAR1B
Host Species Sheep
Immunogen E. coli-derived recombinant human PKA RI beta Met1-Val381 Accession # P31321
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5575
Test Specificity Detects human, mouse, and rat PKA RI beta in Western blots. In Western blots, less than 1% cross-reactivity with recombinant human (rh) PKA RI alpha or rhPKA RII beta is observed; cross-reactivity with PKA RII alpha is unknown.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Human, Mouse, Rat
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.