Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human/Mouse/Rat PKLR Antibody, R&D Systems™

Rabbit Polyclonal Antibody
Supplier: R&D Systems AF8519
This item is not returnable.
View return policy
Description
PKLR Polyclonal specifically detects PKLR in Human, Mouse, Rat samples. It is validated for Western Blot.Specifications
PKLR | |
Polyclonal | |
Western Blot 1:1000 dilution, Simple Western 0.5 μg/mL | |
P30613 | |
PKLR | |
E. coli-derived recombinant human PKLR Leu47-Ser574 Accession # P30613 | |
100 μL | |
Primary | |
Detects human, mouse, and rat PKLR in direct ELISAs and Western blots. In direct ELISAs, less than 1% cross-reactivity with recombinant human PKM-2 is observed. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC, as supplied. 1 month, 2 to 8 degreesC under sterile conditions after opening. 6 months, -20 to -70 degreesC under sterile conditions after opening. | |
IgG |
Western Blot | |
Unconjugated | |
Supplied as a solution in PBS containing BSA, Glycerol and Sodium Azide. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with Sodium Azide | |
EC 2.7.1.40, PK1, PK1PKRL, PKL, PKR, Pyruvate kinase 1, pyruvate kinase isozyme R/L, pyruvate kinase isozymes R/L, pyruvate kinase type L, pyruvate kinase, liver and blood cell, pyruvate kinase, liver and RBC, Red cell/liver pyruvate kinase, RPK, R-type/L-type pyruvate kinase | |
Rabbit | |
Antigen Affinity-purified | |
RUO | |
5313 | |
Human, Mouse, Rat | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction