Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human/Mouse/Rat PKM1/2 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems MAB72442100
This item is not returnable.
View return policy
Description
PKM2 Monoclonal specifically detects PKM2 in Human, Mouse, Rat samples. It is validated for Western Blot.Specifications
PKM2 | |
Monoclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
CTHBP, Cytosolic thyroid hormone-binding protein, MGC3932, OIP3, OIP-3, OIP3EC 2.7.1.40, Opa-interacting protein 3, p58, PK, muscle type, PK2, PK3, PK3PKM, Pyruvate kinase 2/3, pyruvate kinase isozymes M1/M2, Pyruvate kinase muscle isozyme, pyruvate kinase, muscle, TCB, THBP1, THBP1p58, Thyroid hormone-binding protein 1, thyroid hormone-binding protein, cytosolic, Tumor M2-PK | |
E.coli-derived recombinant human PKM2, Ser434-Pro531, Accession No. P14618 | |
100 μg | |
5315.0 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. | |
IgG2b |
Western Blot | |
945131 | |
Western Blot 0.5 μg;/mL | |
P14618 | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
Primary | |
Detects human PKM2 in direct ELISAs. Detects human, mouse, and rat PKM2 in Western blots. | |
Human, Mouse, Rat | |
Supernatant |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction