Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human/Mouse/Rat PKM1/2 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF7244
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF7244 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AF7244 Supplier R&D Systems Supplier No. AF7244
Add to Cart
Add to Cart
This item is not returnable. View return policy

Sheep Polyclonal Antibody

PKM2 Polyclonal specifically detects PKM2 in Human, Mouse, Rat samples. It is validated for Western Blot, Simple Western, Immunohistochemistry, Immunocytochemistry, Knockout Validated.

Specifications

Antigen PKM2
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, KnockDown
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 0.2 ug/mL, Simple Western 2 ug/mL, Immunohistochemistry 5-15 ug/mL, Immunocytochemistry 5-15 ug/mL, Knockout Validated
Gene Accession No. P14618
Gene Alias CTHBP, Cytosolic thyroid hormone-binding protein, MGC3932, OIP3, OIP-3, OIP3EC 2.7.1.40, Opa-interacting protein 3, p58, PK, muscle type, PK2, PK3, PK3PKM, Pyruvate kinase 2/3, pyruvate kinase isozymes M1/M2, Pyruvate kinase muscle isozyme, pyruvate kinase, muscle, TCB, THBP1, THBP1p58, Thyroid hormone-binding protein 1, thyroid hormone-binding protein, cytosolic, Tumor M2-PK
Gene Symbols PKM
Host Species Sheep
Immunogen E. coli-derived recombinant human PKM2 Ser434-Pro531 Accession # P14618
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5315
Test Specificity Detects human, mouse, and rat PKM2 in Western blots and detects recombinant human PKM2 in direct ELISAs.
Reconstitution Sterile PBS to a final concentration of 0.2 mg/mL.
Target Species Human, Mouse, Rat
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.