Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human/Mouse/Rat PKN2 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems MAB5686SP
This item is not returnable.
View return policy
Description
PKN2 Monoclonal specifically detects PKN2 in Human, Mouse, Rat samples. It is validated for Western Blot.Specifications
PKN2 | |
Monoclonal | |
LYOPH | |
Western Blot 1 ug/mL | |
Q16513 | |
PKN2 | |
E. coli-derived recombinant human PKN2 Asp18-Asn207 Accession # Q16513 | |
25 μg | |
Primary | |
Detects endogenous human, mouse and rat PKN2 in Western blots. | |
Human, Mouse, Rat | |
Purified |
Western Blot | |
509105 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
cardiolipin-activated protein kinase Pak2, EC 2.7.11, EC 2.7.11.13, Pak-2, PAK2, PRK2, PRK2MGC150606, PRKCL2, PRKCL2PRO2042, PRO2042, Protein kinase C-like 2MGC71074, protein kinase N2, Protein-kinase C-related kinase 2, serine/threonine-protein kinase N2 | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
5586 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction