Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human/Mouse/Rat PLC-beta 4 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF5128
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF5128 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AF5128 Supplier R&D Systems Supplier No. AF5128
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody

PLC-beta 4 Polyclonal specifically detects PLC-beta 4 in Human, Mouse, Rat samples. It is validated for Western Blot.

Specifications

Antigen PLC-beta 4
Applications Western Blot
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL
Gene Accession No. Q15147
Gene Alias 1-phosphatidyl-D-myo-inositol-4,5-bisphosphate, 1-phosphatidylinositol-4,5-bisphosphate phosphodiesterase beta 4, 1-phosphatidylinositol-4,5-bisphosphate phosphodiesterase beta-4, beta 4 (1-Phosphatidylinositol-4,5-BisphosphatePhosphodiesterase Beta 4)), EC 3.1.4.11, FLJ16169, inositoltrisphosphohydrolase, monophosphatidylinositol phosphodiesterase, phosphoinositidase C, phosphoinositide phospholipase C, phospholipase C, beta 4, PLCB4, PLCbeta 4, triphosphoinositide phosphodiesterase
Gene Symbols PLCB4
Host Species Goat
Immunogen E. coli-derived recombinant human PLC-beta 4 Lys75-Gln250 Accession # Q15147
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5332
Test Specificity Detects endogenous human, mouse, and, rat PLC-beta 4 in Western blots.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Human, Mouse, Rat
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.