Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human/Mouse/Rat Plexin A2 APC-conjugated Antibody, R&D Systems™

Goat Polyclonal Antibody
Supplier: R&D Systems FAB5486A
This item is not returnable.
View return policy
Description
Plexin A2 Polyclonal specifically detects Plexin A2 in Human, Mouse, Rat samples. It is validated for Flow Cytometry.Specifications
Plexin A2 | |
Polyclonal | |
Flow Cytometry 10 uL/10^6 cells | |
FLJ11751, FLJ30634, KIAA0463plexin 2, OCT, OCTSemaphorin receptor OCT, plexin A2, PLXN2, PLXN2plexin-A2, PLXNA2, transmembrane protein OCT | |
Goat | |
Affinity Purified | |
RUO | |
5362 | |
Human, Mouse, Rat | |
IgG |
Flow Cytometry | |
APC | |
P70207 | |
PLXNA2 | |
Chinese hamster ovary cell line CHO-derived recombinant mouse Plexin A2 Met35-Pro1237 Accession # P70207 | |
100 Tests | |
Primary | |
Detects mouse and rat Plexin A2 in direct ELISAs and Western blots. In direct ELISAs, less than 1% cross-reactivity with recombinant mouse (rm) Plexin A1, rmPlexin A3, and recombinant human Plexin B1 is observed. | |
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction