Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human/Mouse/Rat PON3 Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 2 publications
Supplier: R&D Systems AF4345SP
This item is not returnable.
View return policy
Description
PON3 Polyclonal specifically detects PON3 in Human, Mouse, Rat samples. It is validated for Western Blot.Specifications
PON3 | |
Polyclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
EC 3.1.1.2, EC 3.1.1.81, EC 3.1.8.1, paraoxanase-3, paraoxonase 3, serum paraoxonase/lactonase 3 | |
Goat | |
Affinity Purified | |
RUO | |
5446 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Western Blot | |
LYOPH | |
Western Blot 0.5 ug/mL | |
Q15166 | |
PON3 | |
E. coli-derived recombinant human PON3 Ala30-Leu354 Accession # Q15166 | |
25 μg | |
Primary | |
Detects endogenous human, mouse, and rat PON3 in Western blots. In Western blots, this antibody did not cross-react with recombinant human PON1 or PON2. | |
Human, Mouse, Rat | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction