Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
R&D Systems™ Human/Mouse/Rat/Porcine/Canine TGF-beta 1 Quantikine ELISA
Click to view available options
Quantity:
1 Kit
1 Pk. (6 Plates)
Specifications
Specifications
Specificity | Natural and recombinant TGF-beta 1. This assay also recognizes human TGF-beta 1.2. |
For Use With (Application) | ELISA |
Gene ID (Entrez) | 7040 |
Quantity | 1 Kit |
Storage Requirements | Store the unopened product at 2°C to 8°C. Do not use past expiration date. |
Synonym | CEDLAP, DPD1, latency-associated peptide, TGF beta, TGF beta1, TGFB, TGFB1, TGFbeta, TGFbeta 1, TGF-beta 1 protein, TGF-beta-1, transforming growth factor beta-1, transforming growth factor, beta 1 |
Target | Human |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction