Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human/Mouse/Rat PP1 Catalytic Subunit Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF3000
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF3000 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AF3000 Supplier R&D Systems Supplier No. AF3000
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody

PP1 Polyclonal specifically detects PP1 in Human, Mouse, Rat samples. It is validated for Western Blot.

Specifications

Antigen PP1
Applications Western Blot
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL
Gene Accession No. P62136
Gene Symbols PPP1CA
Host Species Goat
Immunogen E. coli-derived recombinant human PP1 alpha Catalytic Subunit Lys41-Pro298 Accession # P62136
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5499
Test Specificity Detects endogenous human, mouse and rat PP1 alpha in Western blots. Based on the immunogenÆs homology, the antibody is expected to cross-react with PP1 beta and PP1 gamma .
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Human, Mouse, Rat
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.