Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human/Mouse/Rat PP2A Catalytic Subunit Antibody, R&D Systems™

Rat Monoclonal Antibody
Supplier: R&D Systems MAB1653
This item is not returnable.
View return policy
Description
PP2A alpha Monoclonal specifically detects PP2A alpha in Human, Mouse, Rat samples. It is validated for Western Blot.Specifications
PP2A alpha | |
Monoclonal | |
LYOPH | |
Western Blot 1 ug/mL | |
P67775 | |
PPP2CA | |
E. coli-derived recombinant human PP2A-alpha catalytic subunit Trp209-Leu309 Accession # P67775 | |
100 μg | |
Primary | |
Detects endogenous human, mouse, and rat PP2A in Western blots. Because PP2A-alpha and PP2A-beta catalytic subunits are identical in the region used for the immunogen, it is expected that both isoforms will be detected by the antibody. | |
Human, Mouse, Rat | |
Purified |
Western Blot | |
234529 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
EC 3.1.3.16, PP2A-alpha, PP2Ac, PP2CA, PP2Calpha, PPP2CA, protein phosphatase 2 (formerly 2A), catalytic subunit, alpha isoform, protein phosphatase 2, catalytic subunit, alpha isozyme, protein phosphatase 2A catalytic subunit, alpha isoform, Replication protein C, RP-C, serine/threonine protein phosphatase 2A, catalytic subunit, alpha isoform, serine/threonine-protein phosphatase 2A catalytic subunit alpha isoform | |
Rat | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
5515 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction